Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Mouse anti-Human, Brilliant Violet 421, Clone: D12, BD Optibuild™

Catalog No. BDB742637 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742637 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742637 Supplier BD Biosciences Supplier No. 742637
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The D12 monoclonal antibody specifically binds to CD11b which is also known as Integrin alpha M (IntegrinαM), Mac-1 subunit alpha (Mac-1a) or complement receptor 3 alpha chain (CR3a). CD11b is encoded by ITGAM /(integrin subunit alpha M) and belongs to the integrin alpha subunit gene family. CD11b is expressed as a ñ165-kDa type I transmembrane glycoprotein that associates with the ñ95-kDa integrin β2(CD18) to form the heterodimeric CD11b/CD18 (αM/β2) complex which also known as Mac-1 or CR3. CD11b is expressed on monocytes, dendritic cells, granulocytes, as well as on some T cells, B cells, and NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for multiple ligands including inactivated C3b (iC3b), ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50) or fibrinogen. Mac-1 regulates leucocyte adhesion and migration, as well as phagocytosis of opsonized particles.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b
Classification Monoclonal
Clone D12
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MAC-1A, Mac-1, ITGAM, Integrin alpha M, CR3A, CR-3 alpha, Mo1, SLEB6
Host Species Mouse
Immunogen Human peripheral blood T lymphocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.