Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Mouse anti-Human, BUV395, Clone: D12, BD Optibuild™

Catalog No. BDB742643 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742643 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742643 Supplier BD Biosciences Supplier No. 742643
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The D12 monoclonal antibody specifically binds to CD11b which is also known as Integrin alpha M (IntegrinαM), Mac-1 subunit alpha (Mac-1a) or complement receptor 3 alpha chain (CR3a). CD11b is encoded by ITGAM /(integrin subunit alpha M) and belongs to the integrin alpha subunit gene family. CD11b is expressed as a ñ165-kDa type I transmembrane glycoprotein that associates with the ñ95-kDa integrin β2(CD18) to form the heterodimeric CD11b/CD18 (αM/β2) complex which also known as Mac-1 or CR3. CD11b is expressed on monocytes, dendritic cells, granulocytes, as well as on some T cells, B cells, and NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for multiple ligands including inactivated C3b (iC3b), ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50) or fibrinogen. Mac-1 regulates leucocyte adhesion and migration, as well as phagocytosis of opsonized particles.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b
Classification Monoclonal
Clone D12
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MAC-1A, Mac-1, ITGAM, Integrin alpha M, CR3A, CR-3 alpha, Mo1, SLEB6
Host Species Mouse
Immunogen Human peripheral blood T lymphocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.