Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Mouse, anti-Human, R718, Clone: D12, BD OptiBuild™

Catalog No. BDB752081 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752081 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752081 Supplier BD Biosciences Supplier No. 752081
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The D12 monoclonal antibody specifically binds to CD11b which is also known as Integrin alpha M (Integrin αM), Mac-1 subunit alpha (Mac-1a) or complement receptor 3 alpha chain (CR3a). CD11b is encoded by ITGAM (integrin subunit alpha M) and belongs to the integrin alpha subunit gene family. CD11b is expressed as a ∼165-kDa type I transmembrane glycoprotein that associates with the ∼95-kDa integrin β2 (CD18) to form the heterodimeric CD11b/CD18 (αM/β2) complex which also known as Mac-1 or CR3. CD11b is expressed on monocytes, dendritic cells, granulocytes, as well as on some T cells, B cells, and NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for multiple ligands including inactivated C3b (iC3b), ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50) or fibrinogen. Mac-1 regulates leucocyte adhesion and migration, as well as phagocytosis of opsonized particles. CAUTION Binding of this CD11b antibody depends on the presence of Ca++. EDTA or ACD, as anticoagulant might affect binding. Using heparin as an anticoagulant or removal of the anticoagulant is recommended.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b
Applications Flow Cytometry
Classification Monoclonal
Clone D12
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.