Learn More
CD11b Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BB515, Clone: ICRF44 (also known as 44), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD11b |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ICRF44 (also known as 44) |
| Conjugate | Brilliant Blue 515 (BB515) |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | MAC-1A; Mac-1; ITGAM; Integrin alpha M; CR3A; CR-3 alpha; Mo1; SLEB6 |
| Host Species | Mouse |
| Immunogen | Human monocytes |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.