Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11b Antibody (908649), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB11241F
Description
CD11b Monoclonal specifically detects CD11b in Mouse samples. It is validated for Flow Cytometry.Specifications
| CD11b | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 908649 | |
| Flow Cytometry | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha M beta 2, Phe17-Asn1105 (Integrin alpha M), Gln24-Asn702 (Integrin beta 2), Accession # NP_032427 (Integrin alpha M) & P11835 (Integrin beta 2) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cellular Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 3684 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction