Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Antibody (3.9), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002711AF488
Description
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD11c | |
Monoclonal | |
Alexa Fluor 488 | |
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
CD11c; gp150/95 a subunit | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
3.9 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
ITGAX | |
Rheumatoid synovial fluid cells and fibronectin purified human monocytes | |
0.1 mL | |
Dendritic Cell Markers, Signal Transduction | |
3687 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction