Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Antibody (AP-MAB0806), PE/Atto594, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB11097871PEATT594
Description
CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| CD11c | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| ITGAX | |
| This CD11c antibody was produced from a hybridoma (mouse myeloma fused with spleen cells)from an Armenian Hamster immunized with mouse lymphocytes. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| AP-MAB0806 | |
| Flow Cytometry | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Armenian Hamster | |
| Protein A or G purified | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction