Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Antibody (AP-MAB0814), Alexa Fluor™ 532, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP106651AF532
Description
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
CD11c | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Armenian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
AP-MAB0814 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
Mouse CD11c+ T Cells | |
0.1 mL | |
Dendritic Cell Markers, Signal Transduction | |
3687 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction