Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Antibody (N418), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281066UV
Description
CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CD11c | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
ITGAX | |
Hamster spleen cells that had been primed to mouse spleen dendritic cells | |
0.1 mL | |
Dendritic Cell Markers, Signal Transduction | |
3687 | |
Human, Mouse | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
N418 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
The antibody binds to mouse and human CD11c. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction