Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Antibody (N418), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281066V
Description
CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CD11c | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| ITGAX | |
| Hamster spleen cells that had been primed to mouse spleen dendritic cells | |
| 0.1 mL | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687 | |
| Human, Mouse | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| N418 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody binds to mouse and human CD11c. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction