Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c Hamster anti-Mouse, BV421, Clone: N418, BD Horizon™
Hamster Monoclonal Antibody
$194.80 - $412.00
Specifications
Antigen | CD11c |
---|---|
Clone | N418 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.
CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.Specifications
CD11c | |
0.2mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4 | |
IgG2 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. |
N418 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Hamster | |
Cell Biology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse Dendritic Cells | |
Primary |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title