Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11c (Integrin αX) Armenian Hamster anti-Mouse, FITC, Clone: N418, BD Phrmingen™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 568943
Description
CD11c (Integrin αX) Monoclonal antibody specifically detects CD11c (Integrin αX) in Mouse samples. It is validated for Flow cytometry
Specifications
CD11c (Integrin αX) | |
Monoclonal | |
0.5 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Armenian Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
N418 | |
FITC | |
Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4 | |
Mouse Dendritic Cells | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction