Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c (Integrin αX), Mouse Anti-Human, RB613, Clone: B-ly6, BD Horizon™

Catalog No. BDB571110 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571110 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB571110 Supplier BD Biosciences Supplier No. 571110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD11c (Integrin αX) Monoclonal antibody specifically detects CD11c (Integrin αX) in Human (QC Testing) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c (Integrin αX)
Applications Flow Cytometry
Classification Monoclonal
Clone B-ly6
Conjugate RB613
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6
Host Species Mouse
Immunogen Spleen Cells from Human with Hairy Cell Leukemia
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3687
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.