Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11c Monoclonal Antibody (3.9), Alexa Fluor™ 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 56011641

Encompass_Preferred

 View more versions of this product

Catalog No. 50-112-4673


Only null left
Edge
Add to Cart

Description

Description

Description: The 3.9 monoclonal antibody reacts with human CD11c, the 150 kDa integrin alphaX chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. Applications Reported: This 3.9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3.9 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11c
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P20702
Itgax
Affinity chromatography
RUO
3687
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
3.9
Alexa Fluor 700
Itgax
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.