Learn More
CD11c Mouse anti-Human, Brilliant Violet 421, Clone: S-HCL-3, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744434
Description
The S-HCL-3 monoclonal antibody specifically binds to CD11c which is encoded by ITGAX (Integrin alpha X, complement component 3 receptor 4 subunit). CD11c is a ñ150 kDa type I transmembrane glycoprotein that is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. CD11c is also present on hairy-cell leukemias and on some acute myeloid leukemias (AML). The S-HCL-3 antibody stains sinus histiocytes of lymph node, tonsil, and spleen; macrophages in the cortex and medulla of thymus; Kupffer cells in liver; elongated intertubular cells in kidney; and alveolar macrophages in lung tissue. In malignant tissue, the antibody stains true histiocytic lymphomas; it does not stain carcinomas or T or B lymphomas. CD11c associates with CD18 (ITGB2/Integrin beta-2) to form the CD11c/CD18 heterodimer which is likewise known as Leukocyte adhesion receptor p150,95 or Complement receptor type 4 (CR4). Ligands for this heterodimeric adhesion receptor include fibrinogen, iC3b, CD23, and CD54 (ICAM-1).
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
CD11c | |
S-HCL-3 | |
Brilliant Violet 421 | |
ITGAX, Integrin alpha-X, AlphaX integrin chain, CR4, Leu M5, p150/95 alpha | |
Human Hairy-Cell Leukemia Cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.