Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Mouse anti-Human, Brilliant Violet 510, Clone: S-HCL-3, BD Optibuild™

Catalog No. BDB744435 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744435 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744435 Supplier BD Biosciences Supplier No. 744435
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S-HCL-3 monoclonal antibody specifically binds to CD11c which is encoded by ITGAX (Integrin alpha X, complement component 3 receptor 4 subunit). CD11c is a ñ150 kDa type I transmembrane glycoprotein that is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. CD11c is also present on hairy-cell leukemias and on some acute myeloid leukemias (AML). The S-HCL-3 antibody stains sinus histiocytes of lymph node, tonsil, and spleen; macrophages in the cortex and medulla of thymus; Kupffer cells in liver; elongated intertubular cells in kidney; and alveolar macrophages in lung tissue. In malignant tissue, the antibody stains true histiocytic lymphomas; it does not stain carcinomas or T or B lymphomas. CD11c associates with CD18 (ITGB2/Integrin beta-2) to form the CD11c/CD18 heterodimer which is likewise known as Leukocyte adhesion receptor p150,95 or Complement receptor type 4 (CR4). Ligands for this heterodimeric adhesion receptor include fibrinogen, iC3b, CD23, and CD54 (ICAM-1).

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Classification Monoclonal
Clone S-HCL-3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGAX, Integrin alpha-X, AlphaX integrin chain, CR4, Leu M5, p150/95 alpha
Host Species Mouse
Immunogen Human Hairy-Cell Leukemia Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.