Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c Mouse anti-Human, Brilliant Violet 786, Clone: S-HCL-3, BD Optibuild™

Catalog No. BDB744439 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744439 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744439 Supplier BD Biosciences Supplier No. 744439
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S-HCL-3 monoclonal antibody specifically binds to CD11c which is encoded by ITGAX (Integrin alpha X, complement component 3 receptor 4 subunit). CD11c is a ñ150 kDa type I transmembrane glycoprotein that is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. CD11c is also present on hairy-cell leukemias and on some acute myeloid leukemias (AML). The S-HCL-3 antibody stains sinus histiocytes of lymph node, tonsil, and spleen; macrophages in the cortex and medulla of thymus; Kupffer cells in liver; elongated intertubular cells in kidney; and alveolar macrophages in lung tissue. In malignant tissue, the antibody stains true histiocytic lymphomas; it does not stain carcinomas or T or B lymphomas. CD11c associates with CD18 (ITGB2/Integrin beta-2) to form the CD11c/CD18 heterodimer which is likewise known as Leukocyte adhesion receptor p150,95 or Complement receptor type 4 (CR4). Ligands for this heterodimeric adhesion receptor include fibrinogen, iC3b, CD23, and CD54 (ICAM-1).

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Classification Monoclonal
Clone S-HCL-3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGAX, Integrin alpha-X, AlphaX integrin chain, CR4, Leu M5, p150/95 alpha
Host Species Mouse
Immunogen Human Hairy-Cell Leukemia Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.