Learn More
CD11c Mouse anti-Human , PE-CF594, Clone: BU15 (also known as BU-15; Bu-15), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD11c |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BU15 (also known as BU-15; Bu-15) |
| Conjugate | PE-CF594 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | ITGAX; integrin alpha X; CD11C; p150 95 integrin alpha chain; SLEB6 |
| Host Species | Mouse |
| Immunogen | Human Synovial Fluid Dendritic Cells |
| Quantity | 0.1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.