Learn More
CD11c Mouse anti-Human, PE-Cy5, Clone: B-LY6, BD
Mouse Monoclonal Antibody
$154.00 - $367.50
Specifications
| Antigen | CD11c |
|---|---|
| Clone | B-LY6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE-Cyanine5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB561692
|
BD Biosciences
561692 |
25 Tests |
Each for $154.00
|
|
|||||
|
BDB551077
|
BD Biosciences
551077 |
100 Tests |
Each for $367.50
|
|
|||||
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
Specifications
| CD11c | |
| Flow Cytometry | |
| PE-Cyanine5 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6 |
| B-LY6 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Spleen Cells from Human with Hairy Cell Leukemia | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.