Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11d/Integrin alpha D Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CD11d/Integrin alpha D |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6028
![]() ![]() |
Proteintech Group Inc
551621AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to CD11d.
Integrin alpha-D/beta-2 is a receptor for ICAM3 and VCAM1. May play a role in the atherosclerotic process such as clearing lipoproteins from plaques and in phagocytosis of blood-borne pathogens, particulate matter, and senescent erythrocytes from the blood.Specifications
CD11d/Integrin alpha D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q13349 | |
3681 | |
Peptide | |
Primary | |
-20°C | |
ITGAD |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADB2, CD11d, Integrin alpha D, integrin, alpha D, ITGAD, Leukointegrin alpha D | |
ITGAD | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title