Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11d Monoclonal Antibody (CA11.8H2)
GREENER_CHOICE

Catalog No. PIMA528277
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528277 Supplier Invitrogen™ Supplier No. MA528277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Dog CD11d antibody, clone CA11.8H2 recognizes the canine CD11d cell surface antigen, also known as integrin alpha d subunit.

Integrin alpha-D/beta-2 is a receptor for ICAM3 and VCAM1. May play a role in the atherosclerotic process such as clearing lipoproteins from plaques and in phagocytosis of blood-borne pathogens, particulate matter, and senescent erythrocytes from the blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11d
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone CA11.8H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene ITGAD
Gene Accession No. 0
Gene Alias ADB2; beta-2 integrin alphaD subunit; CD11 antigen-like family member D; CD11d; FLJ39841; integrin alpha X (Cd11c); integrin alpha-D; integrin subunit alpha D; integrin, alpha D; ITGAD; Itgax; Leukointegrin alpha D
Gene Symbols ITGAD
Host Species Mouse
Immunogen Affinity purified CD11 antigen from canine spleen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 489930
Target Species Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.