Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11R1 Monoclonal Antibody (MIL4)
GREENER_CHOICE

Catalog No. PIMA528278
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528278 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528278 Supplier Invitrogen™ Supplier No. MA528278
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes the porcine cell surface antigen classified as CD11R1.

CD11R1 (ITGAM) may be involved in systemic lupus erythematosus 6 (SLEB6). A chronic, relapsing, inflammatory, and often febrile multisystemic disorder of connective tissue, characterized principally by involvement of the skin, joints, kidneys and serosal membranes. It is of unknown etiology, but is thought to represent a failure of the regulatory mechanisms of the autoimmune system. The disease is marked by a wide range of system dysfunctions, an elevated erythrocyte sedimentation rate, and the formation of LE cells in the blood or bone marrow. Disease susceptibility may be associated with variations affecting the gene represented in this entry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11R1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone MIL4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene ITGAM
Gene Accession No. 0
Gene Alias CD11b; integrin alpha-M; integrin subunit alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); ITGAM
Gene Symbols ITGAM
Host Species Mouse
Immunogen Porcine Lamina Propria Leucocytes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397459
Target Species Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.