Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD11R3 Monoclonal Antibody (2F4/11), FITC
GREENER_CHOICE

Catalog No. PIMA528279
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528279 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528279 Supplier Invitrogen™ Supplier No. MA528279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Pig CD11R3, clone 2F4/11 recognizes porcine CD11R3, a approximately 155 kDa cell surface glycoprotein, member of the alpha integrin family. Mouse anti Pig CD11R3, clone 2F4/11 was clustered as CD11R3 at the Third International Workshop on Swine Leukocyte Differentiation Antigens (Haverson et al. 2001).

CD11R3 is a 155 kDa cell surface glycoprotein, a member of the alpha integrin family. CD11R3 has a similar expression pattern to the human CD11b marker, being expressed on granulocytes, monocytes and alveolar macrophages, but not on lymphocytes, erythrocytes or platelets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11R3
Applications Flow Cytometry
Classification Monoclonal
Clone 2F4/11
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene ITGAM
Gene Accession No. 0
Gene Alias CD11B; integrin alpha-M; integrin subunit alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); ITGAM
Gene Symbols ITGAM
Host Species Mouse
Immunogen Porcine alveolar macrophages.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397459
Target Species Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.