Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 (IL-2R beta) Monoclonal Antibody (TM-B1), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™
SDP

Catalog No. 502544221
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4221 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4221 Supplier Leinco Technologies Inc Supplier No. C63255MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: Clone TM-Beta1 recognizes an epitope on mouse CD122. Endotoxin Level: <0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CD122 (IL-2R beta) Monoclonal antibody specifically detects CD122 (IL-2R beta) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Western Blot

Specifications

Antigen CD122 (IL-2R beta)
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Inhibition Assays, Western Blot
Classification Monoclonal
Clone TM-B1
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Il2rb
Gene Accession No. P16297
Gene Alias CD122; CD122 antigen; high affinity IL-2 receptor beta subunit; high affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL15RB; IL15Rbeta; IL-15Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15Rbeta; IL-2R subunit beta; IL2RB; IL-2RB; IL2RBC; Il-2Rbeta; interleukin 15 receptor, beta; interleukin 2 receptor subunit beta; interleukin 2 receptor, beta; interleukin 2 receptor, beta chain; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70; p70-75; p75
Host Species Rat
Immunogen Rat T-cell line expressing Mouse IL-2RB
Purification Method Protein A/G
Quantity 5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16185
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.