Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD122 Monoclonal Antibody (D4), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $429.50
Specifications
Antigen | CD122 |
---|---|
Clone | D4 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA550851
|
Invitrogen™
MA550851 |
200 μL |
Each for $429.50
|
|
|||||
FEMA550852
|
Invitrogen™
MA550852 |
20 μL |
Each for $195.80
|
|
|||||
Description
CD122 (Interleukin 2 receptor) is involved in T cell-mediated immune responses, primarily expressed in the hematopoietic system and is present in 3 forms. The low affinity form of CD122 is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from CD122. The protein encoded by the CD122 gene represents the beta subunit and is a type I membrane protein. Diseases associate with CD122 protein dysfunction include oligoarticular juvenile idiopathic arthritis and rheumatoid factor negative juvenile idiopathic arthritis.Specifications
CD122 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
CD122; CD122 antigen; high affinity IL-2 receptor beta subunit; high affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL15RB; IL15Rbeta; IL-15Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15Rbeta; IL-2R subunit beta; IL2RB; IL-2RB; IL2RBC; Il-2Rbeta; interleukin 15 receptor, beta; interleukin 2 receptor subunit beta; interleukin 2 receptor, beta; interleukin 2 receptor, beta chain; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70; p70-75; p75 | |
Il2rb | |
IgG2b κ | |
Protein A/G | |
Il2rb |
D4 | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P14784 | |
3560 | |
Recombinant Human IL2Rb protein, Ala27-Thr240 (Accession #P14784). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title