Learn More
CD122 Monoclonal Antibody (TU27), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$172.00 - $419.00
Specifications
Antigen | CD122 |
---|---|
Clone | TU27 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-113-0778
![]() |
Invitrogen™
46122841 |
25 Tests |
Each for $172.00
|
|
|||||
50-113-0779
![]() |
Invitrogen™
46122842 |
100 Tests |
Each for $419.00
|
![]() |
|||||
Description
The interleukin 2 receptor, which is involved in T cell-mediated immune responses, is present in 3 forms with respect to ability to bind interleukin 2. The low affinity form is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from interleukin 2. The protein encoded by this gene represents the beta subunit and is a type I membrane protein.
CD122 (Interleukin 2 receptor) is involved in T cell-mediated immune responses, primarily expressed in the hematopoietic system and is present in 3 forms. The low affinity form of CD122 is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from CD122. The protein encoded by the CD122 gene represents the beta subunit and is a type I membrane protein. Diseases associate with CD122 protein dysfunction include oligoarticular juvenile idiopathic arthritis and rheumatoid factor negative juvenile idiopathic arthritis.Specifications
CD122 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD122; CD122 antigen; high affinity IL-2 receptor beta subunit; high affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL15RB; IL15Rbeta; IL-15Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15Rbeta; IL-2R subunit beta; IL2RB; IL-2RB; IL2RBC; Il-2Rbeta; interleukin 15 receptor, beta; interleukin 2 receptor subunit beta; interleukin 2 receptor, beta; interleukin 2 receptor, beta chain; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70; p70-75; p75 | |
Il2rb | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Il2rb |
TU27 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
P14784 | |
3560 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.