Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 Mouse, anti-Mouse, R718, Clone: TM-β 1, BD OptiBuild™

Catalog No. BDB751980 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751980 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751980 Supplier BD Biosciences Supplier No. 751980
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TM-β1 monoclonal antibody specifically recognizes the 90-100-kDa β chain shared by the IL-2 and IL-15 receptors (IL-2Rβ, CD122). In the periphery, CD122 is expressed on CD8+ T lymphocytes, NK cells, NK-T cells, dendritic epidermal T cells, subsets of intraepithelial lymphocytes, and macrophages. Small subsets of fetal and adult thymocytes constitutively express CD122. CD122+ cells in the bone marrow include committed NK-cell progenitors. IL-2Rβ expression is upregulated by IL-2. CD122 is a transmembrane glycoprotein of the hematopoietin receptor superfamily which can combine with CD132 (γc) alone or CD132 plus CD25 (IL-2Rα) to form intermediate or high-affinity IL-2 receptor complexes, respectively. The β chain of these complexes, CD122, is involved in signal transduction and immunoregulation. The TM-β1 antibody blocks high affinity binding of IL-2 or IL-15 to IL-2Rβ.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD122
Applications Flow Cytometry
Classification Monoclonal
Clone TM-β 1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.