Learn More
Invitrogen™ CD123 Monoclonal Antibody (6H6), Biotin, eBioscience™
Mouse Monoclonal Antibody
$219.00
Specifications
Antigen | CD123 |
---|---|
Clone | 6H6 |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Description
Description: The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70 kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes. Applications Reported: The 6H6 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 6H6 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.Specifications
CD123 | |
0.5 mg/mL | |
Flow Cytometry | |
Biotin | |
Mouse | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
P26951 | |
3563 | |
Primary | |
Affinity chromatography |
6H6 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
IL3RA | |
CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1 | |
IL3RA | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.