Learn More
CD123 Monoclonal Antibody (6H6), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$435.00
Specifications
Antigen | CD123 |
---|---|
Clone | 6H6 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The A7R34 monoclonal antibody reacts with mouse CD127, the high affinity alpha subunit of the mouse IL-7 receptor. IL-7 receptor alpha chain is expressed by immature B cells in the bone marrow, double-negative (CD4-CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+CD8+) thymocytes. In the periphery, mature T cells express CD127 at low level. A7R34 inhibits binding of IL-7 to its receptor and has been used in in vivo and in vitro studies to elucidate the role of IL-7 in T and B cell development and activation. Binding of A7R34 blocks the binding of SB/199, another antibody which recognizes mouse CD127. Applications Reported: This A7R34 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A7R34 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.Specifications
CD123 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P26951 | |
IL3RA | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
IL3RA |
6H6 | |
Flow Cytometry | |
Super Bright 645 | |
Mouse | |
Human | |
3563 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.