Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD123 Mouse anti-Human, BUV615, Clone: 6H6, BD OptiBuild™

Catalog No. BDB751843 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751843 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751843 Supplier BD Biosciences Supplier No. 751843
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD123 Monoclonal antibody specifically detects CD123 in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD123
Applications Flow Cytometry
Classification Monoclonal
Clone 6H6
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu IL-3RALP (CD123) BUV615 6H6 50 μg
Host Species Mouse
Immunogen Human IL3RA Transfected Cell Line
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity IL-3Rα (CD123)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.