Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD123 Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

$293.50 - $565.00

Specifications

Antigen CD123
Clone 001
Concentration 1 mg/mL
Applications ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543438
View Documents
Invitrogen™
MA543438
50 μL
Each for $293.50
Only null left
Add to Cart
 
PIMA543117
GREENER_CHOICE
View Documents
Invitrogen™
MA543117
100 μL
Each for $565.00
Only null left
Add to Cart
 
Description

Description

Specificity: Mouse IL3Ra

IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.
Specifications

Specifications

CD123
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with no preservative
CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1
IL3RA
IgG
Protein A
Antibody
001
ELISA
Unconjugated
Rabbit
Mouse
P26952
16188
Recombinant Mouse IL3Ra protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
IL3RA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.