Learn More
Description
The mIL4R-M1 monoclonal antibody specifically binds to CD124 which is also known as the α subunit of the mouse Interleukin-4 Receptor (IL-4Rα). The mouse IL-4Rα is a 140 kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and nonhematopoietic cells and cell lines. The cell surface IL-4Rα chain binds IL-4 with high affinity and associates with either the common γ chain (IL-4Rα/ãc; aka, type I IL-4R) or the IL-13 receptor alpha subunit (IL-4Rα/IL-13Rα; aka, type II IL-4R complex) to form two distinct types of signal-transducing IL-4R complexes. The type I IL-4 receptor complex specifically binds IL-4 whereas the type II IL-4R binds and transduces signals from either IL-4 or IL-13. The mIL4R-M1 antibody blocks IL-4 binding to cells and is reported to be a potent inhibitor of IL-4's biological activities. The mIL4R-M1 antibody also recognizes naturally-occurring, soluble truncated forms of IL-4Rα (sIL-4R) that result either from enzymatic cleavage of the cell surface extracellular IL-4Rα domain or from differential mRNAsplicing and secretion by cells. These sIL-4R retain their high-affinity ligand binding domain and appear to either enhance or inhibit IL-4-mediated functions depending on the relative local levels of IL-4 and sIL-4R.
Specifications
Specifications
| Antigen | CD124 |
| Classification | Monoclonal |
| Clone | mIL4R-M1 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Blue 700 (BB700) |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Il4r, IL-4Rα, Il4ra, IL-4RA, IL-4R-alpha, IL4-BP, IL-4 Receptor α chain |
| Host Species | Rat |
| Immunogen | Mouse CTLL-19.4 T Cell Line |
| Purification Method | Affinity Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.