Learn More
Invitrogen™ CD126 Monoclonal Antibody (B-R6), eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ BMS135

Description
Description: Recognizes soluble and membranous human IL-6R. This Antibody has been shown to inhibit IL-6 mediated proliferation of XG-1 cells, and partially blocks binding of IL-6 to its receptor. BMS135 is applicable for FACS analysis IL-6 is a multifunctional cytokine involved in the regulation of the immune response, hematopoiesis and acute phase response. It has been recognized to be a member of the alpha-helical cytokine family. IL-6 exerts its action via a cell surface receptor which consists of two subunits, an 80 kDa ligand binding subunit (gp80) of 468 amino acids and a 130 kDa signal transducing protein (gp130) of 896 amino acid residues. Characterisation of the extracellular portion of the 80 kDa IL-6 receptor revealed the existence of a single immunoglobulin-like domain in the NH2-terminal of the extracellular region, which does not contribute to ligand binding. The remainder of the extracellular domain however is essential for low affinity ligand binding, which consecutively triggers the association of the receptor and gp130 thus forming a high affinity binding site for IL-6. A soluble form of the human gp80 protein has been detected in serum and urine samples. This 55 kDa protein representing the extracellular portion of gp80 is generated by shedding, a process that seems to be controlled by protein kinase C. It is still functional, indicating that soluble gp80 plays a biological role in promoting IL-6 activity. So far, the soluble IL-6 receptor is unique in ...
Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
Specifications
CD126 | |
Monoclonal | |
0.1 mg/mL | |
PBS with no preservative | |
P08887 | |
IL6R | |
Rec. Human IL-6R (CD126). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay | |
B-R6 | |
Unconjugated | |
IL6R | |
CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
Mouse | |
Affinity chromatography | |
RUO | |
3570 | |
4°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.