Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD126 Mouse anti-Human, PE, Clone: M5, BD

Catalog No. BDB551850 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551850 100 Tests
BDB561696 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB551850 Supplier BD Biosciences Supplier No. 551850
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M5 monoclonal antibody specifically binds to human CD126 which is also known as the alpha subunit of the human IL-6 Receptor (IL-6Rα). CD126 is an 80 kDa type I transmembrane glycoprotein, also known as gp80 and B cell stimulatory factor-2 (BSF-2) Receptor. The IL-6Rα subunit associates with the 130-160 kDa gp130 subunit (IL-6 Receptor β chain, CD130), that is shared with the receptor complexes for Leukemia Inhibitory Factor (LIF), Ciliary Neurotropic Factor (CNTF), Oncostatin M (OSM), IL-11, Cardiotropin 1 (CT-1) and possibly Neurotrophin-1/B Cell-Stimulating Factor 3 (NNT-1/BSF-3). The IL-6Rα chain binds IL-6 with low affinity; however the association with CD130 stabilizes the IL-6/IL-6Rα complex resulting in the formation of a high affinity ligand-receptor complex. The IL-6Rβ chain mediates signal transduction. CD126 is expressed at high levels by activated and EBV-transformed B cells, plasma cells and myeloma cells and at lower levels by most leucocytes, epithelial cells, fibroblasts, hepatocytes and neural cells. IL-6Rα exists in soluble form in human serum. The serum levels of soluble IL-6Rα appear to elevate in pathological situations such as multiple myeloma, Grave's disease, juvenile chronic arthritis and HIV. The M5 antibody is directed against an epitope not involved in interactions of CD126 with IL-6 or CD130.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD126
Applications Flow Cytometry
Classification Monoclonal
Clone M5
Conjugate PE
Description Interleukin 6 Receptor alpha chain; IL-6R alpha; IL-6Rα
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mixture of U266, XG-1, and BWD41 cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.