Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD127 Monoclonal Antibody (eBioRDR5), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD127 |
---|---|
Clone | eBioRDR5 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H017T03B06A
![]() |
Invitrogen™
H017T03B06A |
100 Tests |
Each for $430.00
|
|
|||||
H017T02B06A
![]() |
Invitrogen™
H017T02B06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow CytometrySpecifications
CD127 | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
P16871 | |
3575 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBioRDR5 | |
Flow Cytometry | |
NovaFluor Blue 610-70S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R α; IL 7Ra; IL 7Rα; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R α; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Rα; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
IgG1 κ | |
Affinity chromatography | |
Il7r |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title