Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD127 Rat anti-Mouse, Brilliant Violet 421, Clone: A7R34, BD

Catalog No. BDB566377 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566377 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566377 Supplier BD Biosciences Supplier No. 566377
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The A7R34 monoclonal antibody specifically binds to mouse CD127 which is also known as the Inter ). CD127 associates with CD132 ( ) to form a high-affinity signaling IL-7R complex that mediates the biological functions of IL-7. CD127 can also complex with the Thymic Stromal Lymphopoietin Receptor (TSLPR) subunit to bind and mediate cellular responses to TSLP. CD127 is a 65-75 kDa type-I transmembrane protein that belongs to the hematopoietin receptor and the Ig gene superfamilies. Mice lacking CD127 display profoundly impaired development of the B and T lymphoid cell lineages, but display no obvious nonlymphoid abnormalities. CD127 is expressed on common lymphoid progenitors and early stages of B lineage development in the bone marrow, the earliest thymocyte progenitors, CD4-CD8- double-negative and CD4+ and CD8+ single-positive thymocytes, naοve and memory T lymphocytes, + T cells, thymic NK cells, and bone marrow stromal cells. CD127 is downregulated upon activation of naοve T cells and CD127 re-expression identifies effector cells that subsequently differentiate into memory T cells. The A7R34 antibody blocks IL-7 binding to CD127. This antibody has been used in both in vitro and in vivo studies to define the biological roles played by CD127.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD127
Classification Monoclonal
Clone A7R34
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Interleukin-7 receptor alpha chain, IL-7R alpha, IL7RA, IL-7RA, IL-7Rα, Il7
Host Species Rat
Immunogen Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.