Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD13/Aminopeptidase-N Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1079S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
500 μL
7 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1079S1 500 μL
MS1079S0 100 μL
MS1079S 1 mL
MS1079R7 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1079S1 Supplier Epredia™ Supplier No. MS1079S1

Stain granulocytes, monocytes, mast cells, GM-progenitor cells and more with Epredia™ CD13/Aminopeptidase-N Ab-3, Mouse Monoclonal Antibody.

CD13 (also known as aminopeptidase-N) is present on most cells of myeloid origin including granulocytes, monocytes, mast cells, and GM-progenitor cells. It is also expressed by the majority of AML, CML in myeloid blast crisis, and in a smaller fraction of lymphoid leukemias. CD13 is absent from normal lymphocytes, platelets and erythrocytes. CD13 is also present on fibroblasts, endothelial cells, epithelial cells from renal proximal tubules and intestinal brush border, bone marrow stromal cells, osteoclasts, and cells lining bile canaliculi. CD13 plays a role in metabolism of biologically active peptides, in phagocytosis, and in bactericidal/tumoricidal activities. It also serves as a receptor for human coronaviruses (HCV).

Host Species: Mouse

Clone: 38C12

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: C-terminal half of the extracellular domain

Immunogen: Recombinant protein encoding the C-terminal half of the extracellular domain of human CD13

Molecular Weight: 150kDa

Positive Control: Prostate, liver

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD13 Ab-3
Applications Western Blot
Classification Monoclonal
Clone 38C12
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the C-terminal half of the extracellular domain of human CD13
Purification Method Prostate, liver
Quantity 500 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.