Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD13/APN/ANPEN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983144
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3144 Supplier Bioss Supplier No. BS1383R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.

Specifications

Antigen CD13/APN/ANPEN
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ANPEP
Gene Accession No. P15144
Gene Alias alanyl (membrane) aminopeptidase; Alanyl aminopeptidase; alanyl aminopeptidase, membrane; Aminopeptidase M; Aminopeptidase N; aminopeptidase N/CD13; Anpep; Apm; AP-M; APN; AP-N; bAPN; BOS_20123; CD13; cluster of differentiation antigen 13 (CD13); gp150; h AP-N; hAPN; kidney aminopeptidase M; Kidney Zn peptidase; KZP; Lap1; Lap-1; leucine arylaminopeptidase 1; membrane alanine aminopeptidase; membrane alanyl aminopeptidase; Membrane protein p161; Microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13; P150; PEPN; rAPN
Gene Symbols ANPEP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CD13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.