Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD13 Monoclonal Antibody (WM15)
GREENER_CHOICE

Catalog No. PIMA544064
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544064 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544064 Supplier Invitrogen™ Supplier No. MA544064
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody The antibody WM15 recognizes an extracellular epitope of human CD13 cell surface glycoprotein, a 150 kDa molecule expressed on granulocytes, endothelial cells, epithelial cells and myeloid progenitors. Functional application: The antibody inhibits infection of cells by human coronavirus and inhibits aminopeptidase N activity of the CD13 molecule immunoprecipitants.

CD13, also known as ANPEP, is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on a wide variety of cell types and tissues. It is prominently found on monocytes and granulocytes in the blood, but not on lymphocytes. Additionally, CD13 is expressed on endothelial cells, epithelial cells, fibroblasts, kidney proximal tubules, intestine, placenta, and myeloid leukemia cells. As a metalloprotease, CD13 preferentially catalyzes the removal of neutral amino acids from small peptides, thereby activating or inactivating bioactive peptides. It plays a crucial role in various biological processes, including the degradation of enkephalins and endorphins, terminal degradation of peptides, amino acid scavenging, antigen processing, and the adhesion and migration of cells. CD13 is also involved in extracellular matrix degradation, signal transduction, inflammatory responses, regulation of intercellular contact, cell motility, and vascularization. In the context of disease, CD13 is associated with the protection of leukemic cells against apoptosis and is linked to a poor prognosis in carcinomas. Furthermore, CD13 functions as an aminopeptidase enzyme and serves as an alpha receptor for coronavirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD13
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone WM15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ANPEP
Gene Accession No. P15144
Gene Alias alanyl (membrane) aminopeptidase; alanyl (membrane) aminopeptidase (aminopeptidase N, aminopeptidase M, microsomal aminopeptidase, CD13, p150); alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase; aminopeptidase Ey; Aminopeptidase M; aminopeptidase N; aminopeptidase N.; aminopeptidase N/CD13; Anpep; APDE; Apm; AP-M; APN; AP-N; bAPN; cAPN; Cd13; cluster of differentiation antigen 13 (CD13); fAPN; gp130; gp150; h AP-N; hAPN; kidney aminopeptidase M; kidney Zn peptidase; KZP; Lap1; Lap-1; leucine arylaminopeptidase 1; LOW QUALITY PROTEIN: aminopeptidase N; membrane alanine aminiopeptidase; membrane alanine aminopeptidase; membrane alanyl aminopeptidase; membrane protein p161; Microsomal aminopeptidase; myeloid plasma membrane glycoprotein CD13; p150; pAPN; PEPN; porcine aminopeptidase N; rAPN; rbAPN; Unknown (protein for MGC:126949)
Gene Symbols ANPEP
Host Species Mouse
Immunogen Human AML cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290
Target Species Human, Monkey
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.