Learn More
CD13 Mouse anti-Human, PE, Clone: WM15, BD
Mouse Monoclonal Antibody
$116.50 - $278.00
Specifications
Antigen | CD13 |
---|---|
Clone | WM15 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560998
|
BD Biosciences
560998 |
25 Tests |
Each for $116.50
|
|
|||||
BDB555394
|
BD Biosciences
555394 |
100 Tests |
Each for $278.00
|
|
|||||
Description
The WM15 monoclonal antibody specifically binds to the 150 kDa Type II integral membrane glycoprotein aminopeptidase N. This antibody binds to GM-progenitor cells, granulocytic and monocytic cells, and mast cells, but not to lymphocytes, platelets or erythrocytes. Aminopeptidase N is involved in the metabolism of many regulatory peptides.
Specifications
CD13 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
WM15 | |
Monoclonal | |
Mouse | |
Human | |
Human AML Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.