Learn More
CD13 Mouse, anti-Human, R718, Clone: L138, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752134
Description
The L138 monoclonal antibody (also known as Leu-M7) specifically binds to a glycosylated 150 kDa type II integral membrane zinc-metalloprotease. The CD13 antigen is also known as aminopeptidase N, APN, ANPEP, and gp150. The CD13 antigen is expressed on granulocytes, monocytes, mast cells, and granulocyte/macrophage progenitor cells (CFU-GM), but not on lymphocytes, platelets, or erythrocytes. It is expressed on most acute myeloid leukemia (AML) cells and some chronic myeloid leukemia (CML) cells. The CD13 antigen is also expressed on epithelial cells of the kidney, small intestine, and respiratory tract, as well as in synaptic membranes in the central nervous system (CNS). The CD13 antigen is involved in the metabolism of many regulatory peptides. Clustering of the CD13 antigen by various forms of ligation promotes the adhesion between monocytes and endothelial cells. The CD13 antigen is the receptor for human coronavirus 229E, the causative agent for some cases of upper respiratory infection.
Specifications
| CD13 | |
| Monoclonal | |
| R718 | |
| 50 μg | |
| Primary | |
| Refrigerate Upon Arrival |
| Flow Cytometry | |
| L138 | |
| Mouse | |
| RUO | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.