Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD13 Mouse, anti-Mouse, R718, Clone: R3-242, BD Horizon™

Catalog No. BDB566982 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566982 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566982 Supplier BD Biosciences Supplier No. 566982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The R3-242 monoclonal antibody specifically binds to aminopeptidase N (APN), which has been reported to be the mouse CD13 homologue. APN is expressed in a variety of mammalian tissues, including intestinal and renal epithelial brush border, endothelium, liver, lung, and brain. In lymphoid tissues, R3-242 mAb is reported to detect APN on monocytes, macrophages, mast cells, dendritic cells, and B lymphocytes, but not on T cells, and thymocytes. However, the reported reactivity with B cells may be non-specific, via binding to the Fcγ receptors (CD16/CD32). This observation is in agreement with reports using different antibodies to mouse APN. APN may play a role in antigen presentation in the immune system. R3-242 mAb does not inhibit the enzymatic activity of APN.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD13
Applications Flow Cytometry
Classification Monoclonal
Clone R3-242
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.