Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586580
Description
Purity is > 95% by SDS-PAGE.
CD13 (ANPEP) is a 150-170 kDa type II transmembrane zinc-binding ectopeptidase expressed on various cell types. CD13 is a metalloprotease that preferentially catalyzes removal of neutral amino acids from small peptides, thus activating or inactivating bioactive peptides. CD13 also has a role in extracellular matrix degradation, antigen processing and signal transduction, is important in inflammatory responses, regulates intercellular contact, cell motility and vascularization. CD13 is expressed by granulocytes, monocytes, myeloid leukemia cells, and is involved in protection of leukemic cells against apoptosis and its expression associated with poor prognosis of carcinomas. CD13 functions as an aminopeptidase enzyme and is also alpha receptor for coronavirus.
Specifications
CD13 | |
Polyclonal | |
Unconjugated | |
ANPEP | |
alanyl (membrane) aminopeptidase; alanyl (membrane) aminopeptidase (aminopeptidase N, aminopeptidase M, microsomal aminopeptidase, CD13, p150); alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase; aminopeptidase Ey; Aminopeptidase M; aminopeptidase N; aminopeptidase N.; aminopeptidase N/CD13; Anpep; APDE; Apm; AP-M; APN; AP-N; bAPN; cAPN; Cd13; cluster of differentiation antigen 13 (CD13); fAPN; gp130; gp150; h AP-N; hAPN; kidney aminopeptidase M; kidney Zn peptidase; KZP; Lap1; Lap-1; leucine arylaminopeptidase 1; LOW QUALITY PROTEIN: aminopeptidase N; membrane alanine aminiopeptidase; membrane alanine aminopeptidase; membrane alanyl aminopeptidase; membrane protein p161; Microsomal aminopeptidase; myeloid plasma membrane glycoprotein CD13; p150; pAPN; PEPN; porcine aminopeptidase N; rAPN; rbAPN; Unknown (protein for MGC:126949) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16790, 290, 81641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P15144, P15684, P97449 | |
ANPEP | |
Synthetic peptide, corresponding to amino acids 880-930 of Human CD13. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction