Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD131 Monoclonal Antibody (1D5E4)
GREENER_CHOICE

Catalog No. FERMA548430
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548430 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548430 Supplier Invitrogen™ Supplier No. MA548430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD131 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD131
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 1D5E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Csf2rb
Gene Accession No. P32927
Gene Alias Ai2ca; AI848964; AIC2A; Aic2b; Bc; beta c; beta common cytokine receptor; BetaIl3; Bil3; CD antigen CD131; CD131; CD131 antigen; CDw131; colony stimulating factor 2 receptor beta common subunit; colony stimulating factor 2 receptor, beta 1, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta 2, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); colony-stimulating factor 2 receptor subunit beta-2; colony-stimulating factor-2 receptor, beta, low-affinity; common beta chain; CSF2RB; Csf2rb1; Csf2rb2; Csfgmrb; cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; IL-3 RB; IL-3 Receptor beta; IL-3 receptor class 2 subunit beta; Il3r; IL3R beta; IL-3R class 2 subunit beta; IL3RB; Il3rb1; Il3rb2; IL5RB; interleukin 3 common beta chain; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, beta (high affinity); interleukin-3 receptor class 2 subunit beta; Interleukin-3 receptor class II beta chain; SMDP5
Gene Symbols Csf2rb
Host Species Mouse
Immunogen Purified recombinant fragment of human CD131 (AA: extra 17-149) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.