Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD131 Mouse anti-Human, Brilliant Violet 650, Clone: 3D7, BD Optibuild™

Catalog No. BDB743319 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743319 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743319 Supplier BD Biosciences Supplier No. 743319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3D7 antibody reacts with CD131, the 120 kD commonβchain (βc) which is shared with the receptor complexes for human granulocyte-macrophage colony stimulating factor (GM-CSFR), interleukin-3 (IL-3R) and interleukin-5 (IL-5R). Together with theαsubunit of either the IL-3R (IL-3Rα), IL-5R (IL-5Rα), or GM- CSFR (GM-CSFRα), the commonβchain forms high-affinity, signaling receptors for human IL-3, IL-5 and GM-CSF, respectively. Cell surfaceβc are expressed by a variety of different cell types including hematopoietic progenitor cells derived from pluripotent stem cells, monocytes, neutrophils, eosinophils, basophils, endothelial cells, fibroblasts, and Langerhans cells. The immunogen used to generate this hybridoma was cells co-transfected with expression vectors which contained cDNA for the human IL-3αandβchains.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD131
Classification Monoclonal
Clone 3D7
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CSF2RB, Cytokine receptor common β, βc, CDw131, IL-3Rβ/IL-5Rβ/GM-CSFRβ
Host Species Mouse
Immunogen Human CD131 Transfected COS Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.