Learn More
CD131 Mouse, anti-Human, R718, Clone: 3D7, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752168
Description
The 3D7 antibody reacts with CD131, the 120 kD common β chain (βc) which is shared with the receptor complexes for human granulocyte-macrophage colony stimulating factor (GM-CSFR), interleukin-3 (IL-3R) and interleukin-5 (IL-5R). Together with the α subunit of either the IL-3R (IL-3Rα), IL-5R (IL-5Rα), or GM- CSFR (GM-CSFRα), the common β chain forms high-affinity, signaling receptors for human IL-3, IL-5 and GM-CSF, respectively. Cell surface βc are expressed by a variety of different cell types including hematopoietic progenitor cells derived from pluripotent stem cells, monocytes, neutrophils, eosinophils, basophils, endothelial cells, fibroblasts, and Langerhans cells. The immunogen used to generate this hybridoma was cells co-transfected with expression vectors which contained cDNA for the human IL-3 α and β chains.

Specifications
CD131 | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
3D7 | |
Mouse | |
RUO | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.