Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD132 Monoclonal Antibody (TUGh4), Biotin, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5011750
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-117-50 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-117-50 Supplier Invitrogen™ Supplier No. 13132982
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: This TUGh4 monoclonal antibody reacts with human CD132, the common gamma chain (gamma c) shared by the IL-2, IL-4, IL-7, IL-9, and IL-15 receptors. This approximately 65 kDa receptor component is expressed on T and B lymphocytes, NK cells, monocytes, and granulocytes. CD132 is involved in intracellular signal transduction by associating with effector molecules within the cytoplasm. Mutations in the human gamma chain gene result in X-linked severe combined immunodeficiency (XSCID), a disease characterized by T cell deficiency leading to impairment of cell-mediated and humoral immunity. Applications Reported: This antibody has been reported for use in flow cytometric analysis. Applications Tested: This TUGh4 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD132
Applications Flow Cytometry
Classification Monoclonal
Clone TUGh4
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IL2RG
Gene Accession No. P31785
Gene Alias [g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1
Gene Symbols IL2RG
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3561
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b λ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.