Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Mouse anti-Human, PE, Clone: AG184, BD

Catalog No. BDB555900 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555900 0.2 mg
BDB561699 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555900 Supplier BD Biosciences Supplier No. 555900
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TheAG184 monoclonal antibody specifically binds to the 65-70 kDa common γ subunit (γc) that is shared by the IL-2, IL-4, IL-7, IL-9, IL-15 and IL-21 receptor complexes. The γc receptor is a type 1 transmembrane glycoprotein that is constitutively expressed by most peripheral T and B lymphocytes, NK cells, monocytes and granulocytes. The cytoplasmic domain of the γc chain plays an important role in cytokine-mediated signal transduction. By immunofluorescent staining and flow cytometric analyses, the AG184 antibody has been shown to specifically recognize human γc expressed by cell lines including human γc gene-transfected cell lines which are known to express the human γc chain. The AG184 antibody can bind the γc chain in the receptors complexed with IL-2, IL-4, or IL-7, indicating that the antibody recognizes an epitope which is distinct from the cytokine binding site of the γc chain.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD132
Applications Flow Cytometry
Classification Monoclonal
Clone AG184
Concentration 0.2mg/mL
Conjugate PE
Description IL2RG; IL-2RG; IL-2Rγ; Common gamma chain; γc; CIDX; SCIDX; SCIDX1; IMD4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.