Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD133 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244250AF350
Description
CD133 Polyclonal specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CD133 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PROM1 | |
A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 650-750) [UniProt O54990] | |
0.1 mL | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction