Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD133 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5487767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244250SS 0.025 mg
NBP244250 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP244250SS Supplier Novus Biologicals Supplier No. NBP244250SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CD133 in Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting

CD133 Polyclonal specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD133
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 30% Glycerol with 0.02% Sodium Azide
Gene Alias AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Gene Symbols PROM1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 650-750) [UniProt O54990]
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8842
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.